Orders LinkContactUsLinkComponent
Drum pump sets for solvents
Drum pump sets for solvents
Supplier:   LUTZ PUMPEN
Drum pump sets for solvents
Supplier:  LUTZ PUMPEN
Certificates
Drum pump sets for solvents
Drum pump sets for solvents
2 Options Available Below
Product Details & Documents
Pump tube SS 41-L-SL stainless steel, drum adapter is made with PP. For easely flammable hydrocarbons such as ethanol, gasoline, butanol, iso-propanol, kerosène, methanol and petroleum.
  • Fast assembly
  • Immediately ready for action
  • Ideal for pumping and filling thin bodied liquids
  • Compact compressed air motor powerful and reliable
The explosion proof universal motor ME II 3 is ideally suitable for transferring of many thin bodied easely flammable and combustible liquids.
Pump set consisting of: 1 Motor ME II 3, 1 pump tube SS 41-L-SL stainless steel, 2,5 m solvent hose 1", electrically conductive bound with hose connectors brass, nozzle brass, equipotential bonding cable, drum adapter PP.
 : Explosion proof according to ATEX directive 94/9/EC category 2.
Product Family Options
Product Information
  • Material
    Flow rate
    Immersion depth
    Power
    Availability
    Price
  • 223-0092
    Drum Pump set with motor ME II 3
    Drum Pump set with motor ME II 3
    Stainless steel/PP
    95 l/min
    1000 mm
    460 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 790,00 €
    3 790,00 €
    1 * 1 ST
       3 790,00 €
  • 223-0093
    Drum Pump set with motor ME II 3
    Drum Pump set with motor ME II 3
    Stainless steel/PP
    95 l/min
    1200 mm
    460 W
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 890,00 €
    3 890,00 €
    1 * 1 ST
       3 890,00 €