1,2-Dibromo-4,5-difluorobenzene
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:64695-78-9
:
Formula:C₆H₂Br₂F₂
MDL Number:MFCD00009890
Boiling Pt:208 °C (1013 hPa)
Melting Pt:32.5…33.5 °C
EINECS:265-021-1
UN:1325
ADR:4.1,II
:271.89 g/mol
:GHS02
:GHS07