4-Bromo-2-chlorophenol
Catalog # EHERC10721500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:207.45 g/mol
Boiling Pt:232 °C (1013 hPa)
Melting Pt:50…51 °C
Density:1.617 g/cm³ (20 °C)
EINECS:223-572-5
UN:3082
ADR:9,III
CAS Number:3964-56-5
Formula:C₆H₄BrClO
MDL Number:MFCD00002166