Syringes, Digital Syringes™
Supplier: HAMILTON BONADUZ
Certificates
About this item
The Digital Syringe™ is a calibrated device composed of a serialised syringe and a serialised digital electronic holder.
- Achieve precision up to 0,5%
- Dispense volumes from 500 µl down to 50 nl
- Device also reduces the possibility of plunger bending
A digital readout allows enhanced syringe volume readability, while an adjustable plunger stop assures accurate, reproducible sample injections.
Delivery: Each unit is delivered with a certificate of calibration.