Combined pH Electrodes
About this item
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media. The all-rounder for a large variety of uncritical applications in almost all kinds of industries: Acid content of beverages, acid and base capacity of water samples, pH start applications.
- Integrated glass sleeve for secure fixation in the titration head
- With S7 screw cap and detachable cap or with Plug & Play S12 connection
- Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC/DGi111-SC/DG114-SC/DGi114-SC: ARGENTHAL™ reference system
DG113-SC/DGi113-SC: Ag/AgCl/LiCl reference system
DGi101-SC/DG115-SC/DGi115-SC: ARGENTHAL™ reference system with silver ion trap
DGi116-Solvent: ARGENTHAL™/LiCl reference system
Ordering information: Each electrode is supplied with 25 ml of reference electrolyte, individual quality certificate and unique serial number.
8 Options Available Below
Product Details & Documents
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media. The all-rounder for a large variety of uncritical applications in almost all kinds of industries: Acid content of beverages, acid and base capacity of water samples, pH start applications.
- Integrated glass sleeve for secure fixation in the titration head
- With S7 screw cap and detachable cap or with Plug & Play S12 connection
- Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC/DGi111-SC/DG114-SC/DGi114-SC: ARGENTHAL™ reference system
DG113-SC/DGi113-SC: Ag/AgCl/LiCl reference system
DGi101-SC/DG115-SC/DGi115-SC: ARGENTHAL™ reference system with silver ion trap
DGi116-Solvent: ARGENTHAL™/LiCl reference system
Ordering information: Each electrode is supplied with 25 ml of reference electrolyte, individual quality certificate and unique serial number.
Product Family Options
Product Information
- TypepHTemp.MaterialElectrolyteDiaphr.Ø×LConnectionForAvailabilityPrice
- 662-1894Combined glass pH electrode DGi101-SC, Plug & Play, with sensor chipCombined glass pH electrode DGi101-SC, Plug & Play, with sensor chipDGi101-SC, Plug & Play, micro0 - 140...+100 °CGlass3 mol/L KClCeramic frit3×162,5 mmS12Aqueous media (small samples)Log in for availability1 * 1 ST695,86 €Specifications:
More
Specifications
Cat. No.662-1894 - 662-2159Combined glass pH electrode DGi111-SC, Plug & Play, with sensor chipCombined glass pH electrode DGi111-SC, Plug & Play, with sensor chipDGi111-SC, Plug & Play0 - 140...+80 °CGlass3 mol/L KClCeramic frit12×149,5 mmS12Aqueous mediaLog in for availability1 * 1 ST587,49 €Specifications:
More
Specifications
Cat. No.662-2159 - 662-2478Combined glass pH electrode DGi113-SC, plug & play, with sensor chipCombined glass pH electrode DGi113-SC, plug & play, with sensor chipDGi113-SC, Plug & Play0 - 120...60 °CGlass1 mol/L LiClMovable PTFE sleeve junction12×149,5 mmS12Non aqueous media (acid/base)Log in for availability1 * 1 ST670,00 €Specifications:
More
Specifications
Cat. No.662-2478 - 662-2160Combined glass pH electrode DGi113-SC, Plug & Play, with sensor chipCombined glass pH electrode DGi113-SC, Plug & Play, with sensor chipDGi113-SC, Plug & Play0 - 120...+60 °CGlass1 mol/L LiClMovable PTFE sleeve junction12×149,5 mmS12Non aqueous media (acid/base)Log in for availability1 * 1 ST730,08 €Specifications:
More
Specifications
Cat. No.662-2160 - 662-2475Combined glass pH electrode DGi111-SC, plug & play, with sensor chipCombined glass pH electrode DGi111-SC, plug & play, with sensor chipDGi111-SC, Plug & Play0 - 140...60 °CGlass3 mol/L KClCeramic frit12×149,5 mmS12Aqueous mediaLog in for availability1 * 1 ST550,00 €Specifications:
More
Specifications
Cat. No.662-2475 - 662-2161Combined glass pH electrode DGi114-SC, Plug & Play, with sensor chipCombined glass pH electrode DGi114-SC, Plug & Play, with sensor chipDGi114-SC, Plug & Play0 - 140...+60 °CGlass3 mol/L KClMoveable PTFE sleeve junction12×149,5 mmS12Dirty samples, protein or sulfide containing samplesLog in for availability1 * 1 ST741,49 €Specifications:
More
Specifications
Cat. No.662-2161 - 662-1669Combined glass pH electrode DGi115-SC, Plug & Play, with sensor chipCombined glass pH electrode DGi115-SC, Plug & Play, with sensor chipDGi115-SC, Plug & Play0 - 140...+100 °CGlass3 mol/L KClFixed ground glass sleeve junction12×149,5 mmS12Media with low ion concentration, acetic acids, alcoholsLog in for availability1 * 1 ST625,00 €Specifications:
More
Specifications
Cat. No.662-1669 - 662-2163Combined glass pH electrode DGi116-Solvent, Plug & Play, with sensor chipCombined glass pH electrode DGi116-Solvent, Plug & Play, with sensor chipDGi116-Solvent, Plug & Play0 - 120...+100 °CGlass1 mol/L LiClMovable ground glass sleeve junction12×149,5 mmS12Non aqueous media (acid/base)Log in for availability1 * 1 ST764,30 €Specifications:
More
Specifications
Cat. No.662-2163