Flat flange lids, centre neck, 1 angled side neck
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.