PEEK tubing, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
Polymer tubing, coloured.
- Biocompatible, inert and easy to cut
- Great for high pressure applications
- Many sizes colour-coded for easier inner diameter identification
PEEK (polyetheretherketone) tubing is biocompatible, chemically inert and can be used in place of stainless steel tubing in most liquid analytical systems. Additionally, when the outer diameter is smaller than 1/16”, PEEK tubing can be used in place of more traditional fused silica capillary tubing. PEEK tubing can be used with stainless steel nuts and ferrules, flangeless fittings, Super Flangeless™ fittings, or any Upchurch Scientific-brand universal Fingertight fitting.