



2-Methyl-2-butene 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:513-35-9
:26760-64-5
Formula:C₅H₁₀
MDL Number:MFCD00009276
:70.13 g/mol
Merck Index:12,00649
Boiling Pt:39 °C (1013 hPa)
Melting Pt:–134 °C
Density:0.66 g/cm³
Flash Pt:–45 °C
EINECS:208-156-3
UN:2460
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3860 to 1.3880 (20°C, 589 nm)
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together