Order Entry
Germany
ContactUsLinkComponent
cis-9-Tricosene (muscalure)
cis-9-Tricosene (muscalure)
Catalog # MOLE71199652-1G
Supplier:  Molekula
CAS Number:  
cis-9-Tricosene (muscalure)
Catalog # MOLE71199652-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :322.62 g/mol
Merck Index:12,06288
Boiling Pt:300 °C (1013 hPa)
Density:0.806 g/cm³ (20 °C)
EINECS:248-505-7
CAS Number:27519-02-4
Formula:C₂₃H₄₆
MDL Number:MFCD00008988