Order Entry
Germany
ContactUsLinkComponent
General purpose tape, 924, Scotch® ATG
General purpose tape, 924, Scotch® ATG
Catalog # MMMA9241955
Supplier:  3M
undefined
General purpose tape, 924, Scotch® ATG
Catalog # MMMA9241955
Supplier:  3M

Specifications

  • Width:
    19,0 mm
  • Description:
    Tape, general purpose, consist of acrylic pressure-sensitive adhesive system
  • Pk:
    12 Roll

Specifications

About this item

Acrylic adhesive on a densified kraft paper liner. General purpose adhesive transfer tape.

  • Bond a wide variety of similar and dissimilar materials such as metals, glass, wood, papers, paints, and many plastics

Length: 55,0 m.

Colour: Transparent.