Tetrachloroethylene, EMPLURA®, Supelco®
: Merck
:127-18-4
:
:C₂Cl₄
:MFCD00000834
:165.83 g/mol
:12,09332
:121.1 °C (1013 hPa)
:–22 °C
:1.63 g/cm³ (20 °C)
:1897
:6.1,III
:M-Clarity™ quality level = MQ200
:204-825-9
:GHS08
:GHS09
:Ambient