Petri dishes, disposable
Supplier: Brand
Certificates
About this item
PS, transparent, with lid. Available with or without vent lid. Dishes and lids are assembled robotically to reduce the risk of contamination, in particular bacterial count.
PS, transparent, with lid. Available with or without vent lid. Dishes and lids are assembled robotically to reduce the risk of contamination, in particular bacterial count.