Condensers, coil, double walled
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Coil condensers acc. to DIN 12593 and in addition to DIN, for condensing vapours, with cooling coil and jacket. With NS socket, NS cone and glass olives for water connection.