Orders LinkContactUsLinkComponent
Condensers, Davies
Condensers, Davies
Supplier:   Lenz Laborglas
Condensers, Davies
Supplier:  Lenz Laborglas
Certificates
Condensers, Davies
Condensers, Davies
4 Options Available Below
Product Details & Documents
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two glass olives.
Documents
Product Family Options
Product Information
  • Jacket length
    Cone size
    Socket size
    Availability
    Price
  • 250 mm
    24/29 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 121,00 €
    121,00 €
    1 * 1 ST
       121,00 €
  • 250 mm
    29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €
  • 400 mm
    24/29 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • 400 mm
    29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 147,00 €
    147,00 €
    1 * 1 ST
       147,00 €