Temperature control systems, PRESTO®
About this item
PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exothermic and endothermic reactions with extraordinary speed, they feature high flow rates, constant pressure and a controlled build-up of pump pressure. Changes in the temperature control of liquid viscosity are balanced dynamically. Permanent internal monitoring and self-lubricating pumps contribute to long service life. The integrated touch panel gives the user a clear view of important information. Fingertip control via the touch screen. Multiple interfaces enable remote control of the PRESTO® across networks and in super-ordinated control systems. Wireless remote control of the new PRESTO® can easily be done by using the Wireless TEMP range of products.
- Fingertip control via the touch screen with 14,4 cm colour touch panel
- Up to three user levels with password management
- USB (Host und Device), Ethernet and SD card slot
- Units are whisper-quiet and barely audible when operating
- Filling system is accessible from the top
- Choice of models with environmentally-friendly natural refrigerant
Max. flow rate/max. pressure:
A30: 25 L/min, 0,5 bar
A38/A41/W41: 0 to 50 L/min, 0,1 to 1,6 bar
A40/W40, A80/W80: 16 to 40 L/min, 0,3 to 1,7 bar
A45/A45t/W50/W50t: 35 to 76 L/min, 0,48 to 3,2 bar
A70: 0 to 40 L/min, 0,1 to 1,7 bar W91/W92 (t/tt): 26 to 80 L/min, 0,5 to 3,0 bar W91×/W92× (t/tt): 18 to 70 L/min, 0,8 to 5,5 bar
W93: 0 to 80 L/min, 0,1 to 3 bar W93x: 0 to 70 L/min, 0,1 to 5,5 bar.
: Supplied without accessories. Go to website for details of accessories.
: Cooling capacity at +200, +100, +20, 0, –10, –20, –30 and −40 °C. W91/W92/W93 (t/tt/x): Cooling capacity at +200, +100, +20, 0, –20, –40, –60 and −80 °C.
33 Options Available Below
Product Details & Documents
PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exothermic and endothermic reactions with extraordinary speed, they feature high flow rates, constant pressure and a controlled build-up of pump pressure. Changes in the temperature control of liquid viscosity are balanced dynamically. Permanent internal monitoring and self-lubricating pumps contribute to long service life. The integrated touch panel gives the user a clear view of important information. Fingertip control via the touch screen. Multiple interfaces enable remote control of the PRESTO® across networks and in super-ordinated control systems. Wireless remote control of the new PRESTO® can easily be done by using the Wireless TEMP range of products.
- Fingertip control via the touch screen with 14,4 cm colour touch panel
- Up to three user levels with password management
- USB (Host und Device), Ethernet and SD card slot
- Units are whisper-quiet and barely audible when operating
- Filling system is accessible from the top
- Choice of models with environmentally-friendly natural refrigerant
Max. flow rate/max. pressure:
A30: 25 L/min, 0,5 bar
A38/A41/W41: 0 to 50 L/min, 0,1 to 1,6 bar
A40/W40, A80/W80: 16 to 40 L/min, 0,3 to 1,7 bar
A45/A45t/W50/W50t: 35 to 76 L/min, 0,48 to 3,2 bar
A70: 0 to 40 L/min, 0,1 to 1,7 bar W91/W92 (t/tt): 26 to 80 L/min, 0,5 to 3,0 bar W91×/W92× (t/tt): 18 to 70 L/min, 0,8 to 5,5 bar
W93: 0 to 80 L/min, 0,1 to 3 bar W93x: 0 to 70 L/min, 0,1 to 5,5 bar.
: Supplied without accessories. Go to website for details of accessories.
: Cooling capacity at +200, +100, +20, 0, –10, –20, –30 and −40 °C. W91/W92/W93 (t/tt/x): Cooling capacity at +200, +100, +20, 0, –20, –40, –60 and −80 °C.
Product Family Options
Air cooled models
Product Information
- TypeTemperature RangeTemp. stabilityHeating capacityCooling capacityRefrigerantW×D×HWeightAvailabilityPrice
- A30–30...+250 °C±0,01...0,05 °C2,7 kW0,5; 0,5; 0,5; 0,4; 0,3; 0,2; 0,05; - kWR452A242×583×612 mm62 kgLog in for availability1 * 1 ST11 990,00 €
- A40–40...+250 °C±0,01...0,05 °C2,7 kW1,2; 1,2; 1,2; 0,9; 0,8; 0,6; 0,3; 0,1 kWR449A323×583×662 mm79Log in for availability1 * 1 ST16 600,00 €
- A45–45...+250 °C±0,05...0,1 °C6 kW3,4; 3,4; 3,5; 3,3; 2,6; 1,8; 1,0; 0,3 kWR452A530×665×126 mm210 kgLog in for availability1 * 1 ST29 750,00 €
- A45t–45...+250 °C±0,05...0,1 °C12 kW3,4; 3,4; 3,5; 3,3; 2,6; 1,8; 1,0; 0,3 kWR452A530×665×126 mm210 kgLog in for availability1 * 1 ST32 760,00 €
- A80–80...+250 °C±0,01...0,05 °C1,8 kW1,2; 1,2; 1,2; 1,2; 1,2; 1,1; 1,1; 1,1 kWR507430×650×1258 mm164 kgLog in for availability1 * 1 ST29 020,00 €
- A70–75...+250 °C±0,01…±0,05 °C1,8 kW1; 1; 1; 0,91; -; 0,84; -; 0,75 kWR449A/R170570×745×880 mm138 kgLog in for availability1 * 1 ST24 780,00 €
- A80t–80...+250 °C±0,01...0,05 °C3,4 kW1,2; 1,2; 1,2; 1,2; 1,2; 1,1; 1,1; 1,1 kWR507430×650×1258 mm167 kgLog in for availability1 * 1 ST30 940,00 €
- A85–85...+250 °C±0,05...0,1 °C6 kW2,8; 2,6; 2,5; 2,4; 2,4; 2,4; 2,4; 0,4 kWR507610×845×1250 mm365 kgLog in for availability1 * 1 ST
- A85t–85...+250 °C±0,05...0,1 °C15 kW2,8; 2,6; 2,5; 2,4; 2,4; 2,4; 2,2; 0,4 kWR507610×845×1250 mm365 kgLog in for availability1 * 1 ST
Water cooled models
Product Information
- TypeTemperature RangeTemp. stabilityHeating capacityCooling capacityRefrigerantW×D×HWeightAvailabilityPrice
- W40–40...+250 °C±0,01...0,05 °C2,7 kW1,2; 1,2; 1,2; 1,0; 0,8; 0,55; 0,3; 0,06 kWR449A323×583×662 mm78 kgLog in for availability1 * 1 ST16 980,00 €
- W50–50...+250 °C±0,05...0,1 °C6 kW7; 7; 7,5; 6,5; 5; 3; 1,8; 0,6 kWR452A530×665×1260 mm210 kgLog in for availability1 * 1 ST35 010,00 €
- W50t–50...+250 °C±0,05...0,1 °C12 kW7; 7; 7,5; 6,5; 5; 3; 1,8; 0,6 kWR452A530×665×1260 mm210 kgLog in for availability1 * 1 ST39 030,00 €
- W55-55...+250 °C±0,05...0,1 °C15 kW-; -; 5; 10; 6,5; 4; 2,5; 1,2 kWR452A610×845×1250 mm288 kgLog in for availability1 * 1 ST
- W56–56...+250 °C±0,01...0,05 °C27 kW-; -; 25,6; 23,3; 16,2; 9,1; 5,7; 1,2 kWR449A600×940×1640 mm385 kgLog in for availability1 * 1 ST
- W80–80...+250 °C±0,01...0,05 °C1,8 kW1,2; 1,2; 1,2; 1,2; 1,2; 1,1; 1,1; 1,1 kWR507430×650×1258 mm159 kgLog in for availability1 * 1 ST29 020,00 €
- W80t–80...+250 °C±0,01...0,05 °C3,4 kW1,2; 1,2; 1,2; 1,2; 1,2; 1,1; 1,1; 1,1 kWR507430×650×1258 mm164 kgLog in for availability1 * 1 ST30 940,00 €
- W85–85...+250 °C±0,05...0,1 °C6 kW2,8; 2,6; 2,5; 2,4; 2,4; 2,4; 2,4; 0,4 kWR507610×845×1250 mm335 kgLog in for availability1 * 1 ST
- W85t–85...+250 °C±0,05...0,1 °C15 kW2,8; 2,6; 2,5; 2,4; 2,4; 2,4; 2,2; 0,4 kWR507610×845×1250 mm335 kgLog in for availability1 * 1 ST
Power packages, water cooled models with extended temperature range
Product Information
- TypeTemperature RangeTemp. stabilityHeating capacityCooling capacityRefrigerantW×D×HWeightAvailabilityPrice
- W56x-45...+150 °C±0,05...0,5 °C27 kW-; -; 25,8; 23,1; 16,9; 11,5; 7,1; 3,5 kWR449A600×940×1640 mm385Log in for availability1 * 1 ST
- W91–91...+250 °C±0,05...±0,2 °C18 kW11,0; 11,0; 11,0; 10,0; 9,5; 9,0; 6,5; 1,5 kWR404A/R449A950×1270×1900 mm870 kgLog in for availability1 * 1 ST
- W91tt–91...+250 °C±0,05...±0,2 °C36 kW11,0; 11,0; 11,0; 10,0; 9,5; 9,0; 6,5; 1,5 kWR404A/R449A950×1270×1900 mm870 kgLog in for availability1 * 1 ST
- W91x–91...+250 °C±0,05...±0,2 °C18 kW11,0; 11,0; 11,0; 10,0; 9,5; 9,0; 6,5; 1,5 kWR404A/R449A950×1270×1900 mm870 kgLog in for availability1 * 1 ST
- W91ttx–91...+250 °C±0,05...±0,2 °C36 kW11,0; 11,0; 11,0; 10,0; 9,5; 9,0; 6,5; 1,5 kWR404A/R449A950×1270×1900 mm870 kgLog in for availability1 * 1 ST
- W92–92...+250 °C±0,05...±0,2 °C18 kW29; -; 25; 18; 11; 10; 7; 1,5 kWR404A/R449A950×1270×1900 mm870 kgLog in for availability1 * 1 ST
- W92tt–92...+250 °C±0,05...±0,2 °C36 kW29; -; 25; 18; 11; 10; 7; 1,5 kWR404A/R449A950×1270×1900870 kgLog in for availability1 * 1 ST
- W93–93...+250 °C±0,05…±0,2 °C27 kW19,5; 19,5; 19,5; 19,5; 19,5; 13; 3,5 kWR1270/R170930×1480×1920 mm920 kgLog in for availability1 * 1 ST
- W92x–92...+250 °C±0,05...±0,2 °C18 kW29; -; 25; 18; 11; 10; 7; 1,5 kWR404A/R449A950×1270×1900 mm870 kgLog in for availability1 * 1 ST
- W93x–93...+250 °C±0,05…±0,2 °C27 kW19,5; 19,5; 19,5; 19,5; 19,5; 13; 3,5 kWR1270/R170930×1480×1920930 kgLog in for availability1 * 1 ST
- W92ttx–92...+250 °C±0,05...±0,2 °C36 kW29; -; 25; 18; 11; 10; 7; 1,5 kWR404A/R449A950×1270×1900 mm870 kgLog in for availability1 * 1 ST
Models with natural refrigerant
Product Information
- TypeTemperature RangeTemp. stabilityHeating capacityCooling capacityRefrigerantW×D×HWeightAvailabilityPrice
- A38–45...+250 °C±0,01…±0,05 °C2,7 kW0,79; 0,79; 0,79; 0,73; 0,71; 0,44; 0,28; 0,05 kWR1270330×750×670 mm80 kgLog in for availability1 * 1 ST16 890,00 €
- A41–45...+250 °C±0,01…±0,05 °C2,7 kW1,33; 1,33; 1,33; 1,24; 0,84; 0,46; 0,31; 0,07 kWR1270330×750×67080 kgLog in for availability1 * 1 ST17 400,00 €
- W41–45...+250 °C±0,01…±0,05 °C2,7 kW1,33; 1,33; 1,33; 1,24; 0,84; 0,46; 0,31; 0,07 kWR1270330×750×670 mm83Log in for availability1 * 1 ST18 050,00 €
Models with synthetic refrigerant
Product Information
- TypeTemperature RangeTemp. stabilityHeating capacityCooling capacityRefrigerantW×D×HWeightAvailabilityPrice
- W58x–50...+250 °C±0,05 ... ±0,1 °C27 kW33; 33; 33; 32; 28; 19; 12; 7 kWR449A700×1080×1740 mm486 kgLog in for availability1 * 1 ST
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M16×1 male2× adapters, M24×1,5 female to M16×1 maleJULA8890052All New Presto® series unitsLog in for availability1 * 2 ST - 153,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube ½"2× adapters, M24×1,5 female to tube ½"JULA8890065All New Presto® series unitsLog in for availability1 * 2 ST - 203,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube 1"2× adapters, M24×1,5 female to tube 1"JULA8890066All New Presto® series unitsLog in for availability1 * 2 ST - 203,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M24×1,5 female2× adapters, M24×1,5 female to M24×1,5 femaleJULA8890067All New Presto® series unitsLog in for availability1 * 2 ST - 223,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M30×1,5 male2× adapters, M24×1,5 female to M30×1,5 maleJULA8890068All New Presto® series unitsLog in for availability1 * 2 ST - 246,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 male to M30×1,5 female2× adapters, M24×1,5 male to M30×1,5 femaleJULA8890069All New Presto® series unitsLog in for availability1 * 2 ST - 274,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M30×1,5 female2× adapters, M24×1,5 female to M30×1,5 femaleJULA8890070All New Presto® series unitsLog in for availability1 * 2 ST - 295,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 male to M16×1 female2× adapters, M24×1,5 male to M16×1 femaleJULA8890071All New Presto® series unitsLog in for availability1 * 2 ST - 251,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT ¾" female2× adapters, M30×1,5 female to NPT ¾" femaleJULA8890084All New Presto® series unitsLog in for availability1 * 2 ST - 215,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Profibus DP interface, for instruments with RS232Profibus DP interface, for instruments with RS232JULA8900020All New Presto® series unitsLog in for availability1 * 1 ST - 1 470,00 €
Product Information
- Catalog #ForAvailabilityPrice
- RS485 interface, for instruments with RS232RS485 interface, for instruments with RS232JULA8900024All New Presto® series unitsLog in for availability1 * 1 ST - 1 470,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Pressure sensor, 2 fittings M30×1,5 male (−95 ... +250 °C)Pressure sensor, 2 fittings M30×1,5 male (−95 ... +250 °C)JULA8980772All New Presto® series unitsLog in for availability1 * 1 ST - 576,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Pressure sensor, 2 fittings M38×1,5 male (−95 ... +250 °C)Pressure sensor, 2 fittings M38×1,5 male (−95 ... +250 °C)JULA8980773All New Presto® series unitsLog in for availability1 * 1 ST - 658,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Electronic module with analog connectors (input, output, standby-in)Electronic module with analog connectors (input, output, standby-in)JULA8900105All New Presto® series unitsLog in for availability1 * 1 ST - 550,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Pressure sensor, 2 fittings M24×1,5 male (−95 ... +250 °C)Pressure sensor, 2 fittings M24×1,5 male (−95 ... +250 °C)JULA8980771All New Presto® series unitsLog in for availability1 * 1 ST - 555,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube ¾"2× adapters, M24×1,5 female to tube ¾"JULA8890064All New Presto® series unitsLog in for availability1 * 2 ST - 140,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M38×1,5 male2× adapters, M30×1,5 female to M38×1,5 maleJULA8890080All New Presto® series unitsLog in for availability1 * 2 ST - 250,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 male to M38×1,5 female2× adapters, M30×1,5 male to M38×1,5 femaleJULA8890081All New Presto® series unitsLog in for availability1 * 2 ST - 250,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT ¾" male2× adapters, M30×1,5 female to NPT ¾" maleJULA8890083All New Presto® series unitsLog in for availability1 * 2 ST - 243,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT 1" male2× adapters, M30×1,5 female to NPT 1" maleJULA8890085All New Presto® series unitsLog in for availability1 * 2 ST - 215,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT 1" female2× adapters, M30×1,5 female to NPT 1" femaleJULA8890086All New Presto® series unitsLog in for availability1 * 2 ST - 236,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to tube 1"2× adapters, M30×1,5 female to tube 1"JULA8890087All New Presto® series unitsLog in for availability1 * 2 ST - 190,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube ¼"2× adapters, M24×1,5 female to tube ¼"JULA8890063All New Presto® series unitsLog in for availability1 * 2 ST - 140,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M38×1,5 female2× adapters, M30×1,5 female to M38×1,5 femaleJULA8890082All New Presto® series unitsLog in for availability1 * 2 ST - 280,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Sight glass, −100…+280 °C, PN16/Class 230, 2 fittings M30×1,5 maleSight glass, −100…+280 °C, PN16/Class 230, 2 fittings M30×1,5 maleJULA8970815All New Presto® series unitsLog in for availability1 * 1 ST - 1 450,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT 1" male2× adapters, M24×1,5 female to NPT 1" maleJULA8890062All New Presto® series unitsLog in for availability1 * 2 ST - 214,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Adapter, M24×1,5 male to M24×1,5 maleAdapter, M24×1,5 male to M24×1,5 maleJULA8890110All New Presto® series unitsLog in for availability1 * 1 ST - 82,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¾" male2× adapters, M24×1,5 female to NPT ¾" maleJULA8890061All New Presto® series unitsLog in for availability1 * 2 ST - 214,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ½" male2× adapters, M24×1,5 female to NPT ½" maleJULA8890060All New Presto® series unitsLog in for availability1 * 2 ST - 214,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ³/₈" male2× adapters, M24×1,5 female to NPT ³/₈" maleJULA8890059All New Presto® series unitsLog in for availability1 * 2 ST - 145,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¼" male2× adapters, M24×1,5 female to NPT ¼" maleJULA8890058All New Presto® series unitsLog in for availability1 * 2 ST - 137,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT 1" female2× adapters, M24×1,5 female to NPT 1" femaleJULA8890057All New Presto® series unitsLog in for availability1 * 2 ST - 274,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¾" female2× adapters, M24×1,5 female to NPT ¾" femaleJULA8890056All New Presto® series unitsLog in for availability1 * 2 ST - 243,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ½" female2× adapters, M24×1,5 female to NPT ½" femaleJULA8890055All New Presto® series unitsLog in for availability1 * 2 ST - 217,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ³/₈" female2× adapters, M24×1,5 female to NPT ³/₈" femaleJULA8890054All New Presto® series unitsLog in for availability1 * 2 ST - 217,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¼" female2× adapters, M24×1,5 female to NPT ¼" femaleJULA8890053All New Presto® series unitsLog in for availability1 * 2 ST - 153,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M38×1,5 female2× adapters, M30×1,5 female to M38×1,5 femaleJULA8890089All New Presto® series unitsLog in for availability1 * 2 ST - 411,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M30×1,5 female2× adapters, M30×1,5 female to M30×1,5 femaleJULA8890088All New Presto® series unitsLog in for availability1 * 2 ST - 302,00 €
Product Information
- Catalog #ForAvailabilityPrice
- EtherCAT interface is a protocol converter from EtherCAT to RS232EtherCAT interface is a protocol converter from EtherCAT to RS232JULA8900025All New Presto® series unitsLog in for availability1 * 1 ST - 1 470,00 €