

Allyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:591-87-7
:
Formula:C₅H₈O₂
MDL Number:MFCD00008721
:100.12 g/mol
Merck Index:12,00293
Boiling Pt:103 °C (1013 hPa)
Melting Pt:–96 °C
Density:0.93 g/cm³ (20 °C)
EINECS:209-734-8
UN:2333
ADR:3 (6.1),II
:M-Clarity™ quality level = MQ200
Flash Pt:6 °C
:GHS02
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:28 g/l (20 °C)
- Melting Point:-96 °C
- Molar Mass:100.11 g/mol
- Boiling Point:103 - 104 °C (1013 mbar)
- Vapor Pressure:27.2 hPa (20 °C)
- Flash Point:6 °C
- Refractive Index:1.4042 (20 °C, 589 nm)
- Explosion Limit:2.1 - 13.0 %(V)
- Density:0.93 g/cm3 (20 °C)
- Ignition Point:374 °C
- Synonyms:Acetic acid allyl ester
- Hill Formula:C5H8O2
- Chemical Formula:CH3COOCH2CH=CH2
Specification Test Results
Frequently Bought Together