Order Entry
Germany
ContactUsLinkComponent
 
2,3-Dibromopropene
Catalog # MOLEM26909269
Supplier:  Molekula
CAS Number:  
2,3-Dibromopropene
Catalog # MOLEM26909269
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :199.87 g/mol
Merck Index:13,03050
Boiling Pt:42 °C (1013 hPa)
Density:2.05 g/cm³ (20 °C)
EINECS:208-155-8
 :GHS07
Storage Temperature:Refrigerator
CAS Number:513-31-5
Formula:C₃H₄Br₂
MDL Number:MFCD00000211