



4-Methylheptane, Sigma-Aldrich®
Catalog # SIAL111023-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:114.23 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:–121 °C
Density:0.7046 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-650-1
UN:1262
ADR:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:589-53-7
Formula:C₈H₁₈
MDL Number:MFCD00009409