



3-Methylheptane
Catalog # EHERC15087800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:114.23 g/mol
Boiling Pt:119 °C (1013 hPa)
Melting Pt:–121 °C
Density:0.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-660-6
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:589-81-1
Formula:C₈H₁₈
MDL Number:MFCD00027244