Hand pads, medium, 7440, Scotch-Brite™
Catalog # MMMA07440
Supplier: 3M
Specifications
- Colour:Brown
- L×W:224×158 mm
- Pk:20
Specifications
About this item
Tight and dense, this pad is the most aggressive and durable for quick stock removal. It is excellent for heavy cleaning, deburring, and finishing. May be used by hand, with a hand pad block or on an in-line sander.
- Cuts fast to remove contaminants and oxides in the most demanding cleaning and finishing applications
- Provides similar performance as steel wool grade 4
Abrasive mineral: Medium grade aluminium oxide.