Disposable earplugs, E-A-RSoft™
Supplier: 3M
Certificates
About this item
E-A-RSoft™ yellow neon earplugs are made from a slow expanding polyurethane foam material. A plug that gives evenly distributed pressure allows flexibility and a good seal with optimum comfort. Once inserted in the ear, the foam plugs expand to provide a snug and secure custom fit. These roll down earplugs are available with and without cord and be used with the One Touch™ dispenser.
- Soft hypoallergenic foam material for lower pressure inside the ear
- Smooth, dirt resistant surface for better hygiene, durability and comfort
- Polyester cord helps prevent hearing loss
- One size that fits most ear canals
SNR: 37 dB
Certifications: EN 352-2