


Isobutylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-81-9
:
Formula:C₄H₁₁N
MDL Number:MFCD00008146
:73.14 g/mol
Merck Index:13,05149
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–85 °C
Density:0.7344 g/cm³ (20 °C)
EINECS:201-145-4
UN:1214
ADR:3 (8),II
:M-Clarity™ quality level = MQ200
Flash Pt:–9 °C
:GHS02
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:1000 g/l (25 °C)
- Melting Point:-85 °C
- Molar Mass:73.14 g/mol
- Boiling Point:68 °C (1013 hPa)
- Vapor Pressure:135 hPa (20 °C)
- Flash Point:-10 °C
- Explosion Limit:2 - 9 %(V)
- Density:0.736 g/cm3 (20 °C)
- pH:12.2 (100 g/l, H2O, 20 °C)
- Ignition Point:400 °C