Serological pipettes
Supplier: HIRSCHMANN
Certificates
About this item
Measuring pipettes made of AR-Glas®, class B.
- With silver diffusion ink durable brown graduation
- Wide opening, with expanded tip opening
- Zero point at top, graduated to tip
- With standard cotton plug (pipettes with 5 ml and more)