4-Hydroxy-4-methyl-2-pentanone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-42-2
:
Formula:(CH₃)₂C(OH)CH₂COCH₃
MDL Number:MFCD00004471
:116.16 g/mol
Merck Index:14,2964
Boiling Pt:168 °C
Melting Pt:–47 °C
Density:0.935
Flash Pt:58 °C (136 °F)
Storage Temperature:Ambient
EINECS:204-626-7
UN:1148
ADR:3,III
REACH:01-2119473975-21
:GHS02
:GHS07