
4-Hydroxy-3-methylbutanone tech. 85%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3393-64-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00004739
Boiling Pt:90 °C (1013 hPa)
Density:0.9908 g/cm³ (20 °C)
:102.13 g/mol
EINECS:222-238-6
:GHS07
Storage Temperature:Ambient