Propagyl amine hydrochloride, Sigma-Aldrich®
Catalog # SIALP50919-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:91.54 g/mol
Melting Pt:178…179 °C
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:239-440-5
CAS Number:15430-52-1
Formula:C₃H₆ClN
MDL Number:MFCD00012907