ω,4-Dibromoacetophenone
Marketsource
Supplier: Thermo Scientific
Jump to:
About this item
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:107…108 °C
- Storage Temperature:Refrigerator
- EINECS:202-783-6
- UN:3261
- ADR:8,II
3 Options Available Below
Product Details & Documents
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:107…108 °C
- Storage Temperature:Refrigerator
- EINECS:202-783-6
- UN:3261
- ADR:8,II
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



