0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1-Ethoxy-2,2,2-trifluoroethanol
1-Ethoxy-2,2,2-trifluoroethanol tech. 80%, Technical Grade
1-Ethoxy-2,2,2-trifluoroethanol tech. 80%, Technical Grade
:
Thermo Fisher Scientific
1-Ethoxy-2,2,2-trifluoroethanol tech. 80%, Technical Grade
:
Thermo Fisher Scientific
:
433-27-2
:
:
C₄H₇F₃O₂
:
MFCD00000441
:
144.09 g/mol
:
104…106 °C
:
1.230
:
39 °C (102 °F)
:
Ambient
:
207-087-6
:
1989
:
3,III
:
GHS02
:
GHS07
msds_ALFAL01554
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years