Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 3-(trimethylsilyl)propiolate
Ethyl-3-(trimethylsilyl)propiolate ≥98%
Ethyl-3-(trimethylsilyl)propiolate ≥98%
:
Thermo Fisher Scientific
Ethyl-3-(trimethylsilyl)propiolate ≥98%
:
Thermo Fisher Scientific
:
16205-84-8
:
:
C₈H₁₄O₂Si
:
MFCD00074886
:
170.28 g/mol
:
92…96 °C (28 mmHg)
:
0.897
:
71 °C (160 °F)
:
GHS07
:
Ambient
Thermo Scientific Acroseal 25 Years