1,11-Dodecadiene
Glass bottle, 25 mL
Catalog # ABCRAB135141.0025
Marketsource
Supplier: abcr
Jump to:
Product Details & Documents
Synonyms: Dodeca-1,11-diene
- CAS Number:5876-87-9
- Formula:C₁₂H₂₂
- MDL Number:MFCD00048156
- Molecular Weight:166.31 g/mol
Specifications
Specifications
- Pack type:Glass bottle
- Pk:25 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



