Filter cartridges, Sartopore® 2
Sartofluor® two filter systems are ideally suited for bioburden reduction and particle removal of highly viscous and difficult to filter pharmaceutical solutions. Due to its hydrophilic PES membrane, the filter offers outstanding total throughputs and flow rate performances. Thus ensuring highest process efficiency, minimised filtration costs and short filtration cycle times.
- Compatible to pH-range from 1 to 14
- Ideal to scale-up
Membrane: Hydrophilic PES; support fleece: PP; end cap, core and cage: PP; O-rings: silicone; housing: PP; filling bell: PC
Maximum differential pressure: five bar at 20 °C; two bar at 80 °C