



n-Hexane, Isocratic Grade for HPLC, for spectroscopy
Catalog # KRAF12510.4700
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520