Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Hexane
n-Hexane, extra pure
n-Hexane, extra pure
:
Bernd Kraft
n-Hexane, extra pure
:
Bernd Kraft
:
110-54-3
:
92112-69-1
:
H₃C(CH₂)₄CH₃
:
MFCD00009520
:
86.18 g/mol
:
13,04712
:
69 °C (1013 hPa)
:
–94.3 °C
:
0.659 g/cm³ (20 °C)
:
–22 °C
:
203-777-6
:
1208
:
3,II
:
GHS02
:
GHS08
:
GHS07
:
GHS09
:
Ambient