Order Entry
Germany
ContactUsLinkComponent
n-Hexadecane for synthesis, Sigma-Aldrich®
Health
n-Hexadecane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Health
n-Hexadecane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    18 °C
  • Molar Mass:
    226.44 g/mol
  • Boiling Point:
    287 °C
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    135 °C
  • Density:
    0.77 g/cm3 (20 °C)
  • Viscosity kinematic:
     - 7 mm2/s (40 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number