Order Entry
Germany
ContactUsLinkComponent
 
n-Hexadecane 99%, pure
 
undefined
n-Hexadecane 99%, pure
 

 

  • Appearance:
    Clear colorless Liquid or solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4330 to 1.4350 (20°C, 589 nm)
  • GC:
    ≥98.5 %