Order Entry
Germany
Orders LinkContactUsLinkComponent
n-Eicosane, Supelco®

n-Eicosane, Supelco®

Glass bottle for solids, 5 g
Catalog # 44818-5G
Supplier:   Merck
n-Eicosane, Supelco®
Catalog # 44818-5G
Supplier:  Merck
CAS Number:  112-95-8
n-Eicosane, Supelco®
n-Eicosane, Supelco®

Glass bottle for solids, 5 g

CAS Number:  112-95-8
Price
188,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  N-Eicosane
  • CAS Number:112-95-8
  • Formula:H₃C(CH₂)₁₈CH₃
  • MDL Number:MFCD00009344
  • Molecular Weight:282.55 g/mol
  • Boiling Pt:342.7 °C (1013 hPa)
  • Melting Pt:34…37 °C
  • Density:0.932 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:204-018-1

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications