Order Entry
Germany
Orders LinkContactUsLinkComponent
n-Eicosane ≥99%

n-Eicosane ≥99%

n-Eicosane ≥99%
Certificates
n-Eicosane ≥99%
n-Eicosane ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 112-95-8 Molecular Formula: CH3(CH2)18CH3 Formula Weight: 282.54 Flash Point: 176°(348°F) Refractive Index: 1.4425 Melting Point: 36-38° Boiling Point: 220°/30mm Beilstein Reference: 1700722 MDL No.: MFCD00009344

Synonyms:  N-Eicosane
  • CAS Number:112-95-8
  • Formula:H₃C(CH₂)₁₈CH₃
  • MDL Number:MFCD00009344
  • Molecular Weight:282.55 g/mol
  • Boiling Pt:220 °C (30 mmHg)
  • Melting Pt:36…38 °C
  • Density:0.7888
  • Flash Pt:176 °C (348 °F)
  • Storage Temperature:Ambient
  • EINECS:204-018-1
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.