Order Entry
Germany
ContactUsLinkComponent
Sodium octyl sulphate, Supelco®
Sodium octyl sulphate, Supelco®
Catalog # 75073-10G
Supplier:  Merck
CAS Number:  
undefined
Sodium octyl sulphate, Supelco®
Catalog # 75073-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :232.28 g/mol
Melting Pt:181…183 °C
Density:1.266 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:205-535-5
CAS Number:142-31-4
Formula:C₈H₁₇NaO₄S
MDL Number:MFCD00007470