Sodium dodecyl sulphate (SDS) ≥85%, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
: Merck
:151-21-3
:
:CH₃(CH₂)₁₁OSO₃Na
:MFCD00036175
:288.38 g/mol
:14,08636
:204…207 °C
:1.1 g/cm³ (20 °C)
:>100 °C
:Ambient
:205-788-1
:1325
:4.1,III
:GHS02
:GHS05
:GHS07
- Assay (two-phase titration):≥ 85.0 %
- Identity (IR-spectrum):passes test
- Identity (wet chemistry):passes test
- Appearance:White to pale yellow powder.
- Alkaline reacting substances:passes test
- Unesterified alcohol:≤ 4.0 %
- Sodium chloride + sodium sulfate:≤ 8.0 %
- Cu (Copper)*:≤ 30 ppm
- Ni (Nickel)*:≤ 2 ppm
- V (Vanadium)*:≤ 1 ppm
- Residual solvents (ICH Q3C):excluded by production process