Order Entry
Germany
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS), Millipore®
Sodium dodecyl sulphate (SDS), Millipore®
Catalog # 428023-500
CAS Number:  
undefined
Sodium dodecyl sulphate (SDS), Millipore®
Catalog # 428023-500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
ADR:4.1,II
 :GHS02
 :GHS06
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175