Order Entry
Germany
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥99%, AnalaR NORMAPUR® analytical reagent
Sodium dodecyl sulphate (SDS) ≥99%, AnalaR NORMAPUR® analytical reagent
Catalog # 108073J
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium dodecyl sulphate (SDS) ≥99%, AnalaR NORMAPUR® analytical reagent
Catalog # 108073J
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99 %
  • Assay (C12 compounds)(calc.on anhydrous):
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Insolubility in water:
    Max. 30 ppm
  • Loss on drying (100°C):
    Max. 1.5 %
  • Cl (Chloride):
    Max. 100 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • Cu (Copper):
    Max. 1 ppm
  • Fe (Iron):
    Max. 2 ppm
  • Pb (Lead):
    Max. 5 ppm
  • Absorbance (220-350 nm) (3 %):
    Max. 0.1

Specification Test Results

About this item

 :
 :288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:100 °C (closed cup)
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
ADR:4.1,III
REACH:01-2119489461-32
 :GHS02
 :GHS05
 :GHS07
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175