0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1-Hexanethiol
1-Hexanethiol ≥97%
1-Hexanethiol ≥97%
:
Thermo Fisher Scientific
1-Hexanethiol ≥97%
:
Thermo Fisher Scientific
:
111-31-9
:
:
C₆H₁₄S
:
MFCD00004909
:
150…154 °C
:
–81 °C
:
0.838
:
30 °C (86 °F)
:
Ambient
:
203-857-0
:
6.1,II
:
118.24 g/mol
:
3071
:
GHS06
:
GHS02
msds_ALFAL09659
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years