1-Heptanethiol for synthesis, Sigma-Aldrich®
Catalog # 8.41753.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Solubility:0.009 g/l (20 °C)
- Melting Point:-43 °C
- Molar Mass:132.26 g/mol
- Boiling Point:173 - 175 °C (1013 hPa)
- Vapor Pressure:1.5 hPa (25 °C)
- Flash Point:46 °C
- Refractive Index:1.4505
- Explosion Limit:0.9 %(V)
- Density:0.844 g/cm3 (20 °C)
Specification Test Results
About this item
:
:132.27 g/mol
Boiling Pt:176 °C (1013 hPa)
Melting Pt:–43.4 °C
Density:0.854 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-678-8
UN:3071
ADR:6.1 (3),II
:M-Clarity™ quality level = MQ200
:GHS02
CAS Number:1639-09-4
Formula:C₇H₁₆S
MDL Number:MFCD00004911