Order Entry
Germany
ContactUsLinkComponent
Glycerine
Glycerine
Catalog # LBCL4564012
Supplier:  LABC LABORTECHNIK
CAS Number:  
undefined
Glycerine
Catalog # LBCL4564012
Supplier:  LABC LABORTECHNIK
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
Storage Temperature:Ambient
EINECS:200-289-5
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722