Glycerine, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
Storage Temperature:Ambient
EINECS:200-289-5