
1,4-Butanediol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-63-4
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00002968
:90.12 g/mol
Boiling Pt:229…230 °C
Melting Pt:19…20 °C
Density:1.017
Flash Pt:135 °C (275 °F)
EINECS:203-786-5
:GHS07
Storage Temperature:Ambient