1,3-Propanediol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:504-63-2
:
Formula:HO(CH₂)₃OH
MDL Number:MFCD00002949
:76.1 g/mol
Merck Index:14,9714
Boiling Pt:214 °C
Melting Pt:–26 °C
Density:1.053
Flash Pt:140 °C (174 °F)
Storage Temperature:Ambient
EINECS:207-997-3
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)