Methanol
Marketsource
Supplier: LABC LABORTECHNIK
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
5 Options Available Below
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- LBCL4561201
- LBCL4561203
- LBCL4561200
- LBCL4561293
- LBCL4561225
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.LBCL4561201Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.LBCL4561203Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.LBCL4561200Environmentally PreferableEnvironmental certificationMarketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.LBCL4561293Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.LBCL4561225Environmentally PreferableEnvironmental certificationMarketsource



