

Ethanol absolute ≥99.8% for molecular biology
A3678.1000
:
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- DNases/RNases/Proteases:not detectable
- Assay (GC):min. 99.8%
- Acidic/alkaline react. subst.:max. 0.0002%
- Non-volatile matter:max. 0.001%
- Water (K.F.):max. 0.2%
:
:46.07 g/mol
:12,03806
:78.2 °C
:–114.1 °C
:0.79 g/cm3
:12 °C
:Ambient
:200-578-6
:1170
:3,II
:01-2119457610-43
:GHS02
:GHS07
:64-17-5
:C₂H₅OH
:MFCD00003568