2-Propanol ≥99.0% Ph. Eur., USP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear colourless liquid
- Relative density:0.785 - 0.789
- Residual solvents (CPMP/ICH/283/95):Conforms
- Residues of metal catalysts or reagents:Conforms
- As (Arsenic):≤ 0.1 ppm
- Cd (Cadmium):≤ 0.1 ppm
- Co (Cobalt):≤ 0.1 ppm
- Hg (Mercury):≤ 0.1 ppm
- Ni (Nickel):≤ 0.1 ppm
- Pb (Lead):≤ 0.1 ppm
- V (Vanadium):≤ 0.1 ppm
- Conforms to Ph.Eur./USP:Passes test
- Ph.Eur. specifications: :.
- Identification B [Ph.Eur.]:1.376 - 1.379
- Identification C:Passes test Ph.Eur.
- Appearance test:Passes test Ph.Eur.
- Acidity or alkalinity:Passes test Ph.Eur.
- Absorbance (230 nm) [Ph.Eur.]:≤ 0.30
- Absorbance (250 nm) [Ph.Eur.]:≤ 0.10
- Absorbance (270 nm) [Ph.Eur.]:≤ 0.03
- Absorbance (290 nm) [Ph.Eur.]:≤ 0.02
- Absorbance (310 nm) [Ph.Eur.]:≤ 0.01
- Absorption curve (*):Passes test Ph.Eur.
- Benzene [Ph.Eur.]:≤ 2 ppm
- Total of impur. excl.2-butanol [Ph.Eur.]:≤ 0.3 %
- Peroxides:Passes test Ph.Eur.
- Non-volatile substances [Ph.Eur.]:≤ 20 ppm
- Water [Ph.Eur.]:≤ 0.5 %
- USP specifications::.
- Assay [USP]:≥ 99
- Identification A:Passes test USP
- Identification B (*):Passes test USP
- Methanol [USP] (*):≤ 0.02 %
- Each other indiv.known impurity [USP](*):≤ 0.1 %
- Individual unspecified impurity [USP](*):≤ 0.1 %
- Total impurities [USP] (*):≤ 1.0 %
- Limit of nonvolatile residue [USP]:≤ 0.005 %
- Specific gravity (25°C) [USP]:0.783 - 0.787
- Refractive index (20°C) [USP]:1.376 - 1.378
- Acidity:Passes test USP
- Water determination [USP]:≤ 0.5 %
- (*): Results based on manufacturer:(*)
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 25L Stainlees steel drum (ZPACK12124024) 5L Aluminium bottle for liquids DIN 45 UN X (ZPACK8000430)
Packaging Information
20904_2 PROPANOL PH.EUR_BSE_Rev2
BSE/TSE Certificates
20904_2 PROPANOL PH.EUR_LATEX_REV0
Additional Product Details
spec_VWRC20904.293 spec_VWRC20904.320 spec_VWRC20904.320 spec_VWRC20904.360 spec_VWRC20904.360
Product specification
More